EasySep™ Human CD8+ T Cell Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19053 19053RF
STMC19053EA 966 EUR
STMC19053 STMC19053RF
EasySep™ Human CD8+ T Cell Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human CD8+ T cells.
  • Fast, easy-to-use and column-free
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Human CD8+ T Cell Enrichment Kit is designed to isolate CD8+ T cells from fresh or previously frozen peripheral blood mononuclear cells or ammonium chloride-lysed leukapheresis sample by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-CD8+ T cells and glycophorin A and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells are poured off into a new tube.
For even faster cell isolations, we recommend the new EasySep™ Human CD8+ T Cell Isolation Kit which isolates cells in just 8 minutes.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').