EasySep™ HLA T Cell Enrichment Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19051HLA 19051HLARF
STMC19051HLAEA 1111 EUR
STMC19051HLA STMC19051HLARF
EasySep™ HLA T Cell Enrichment Kits
Media voor celscheiding
Immunomagnetic isolation of untouched human T cells for HLA analysis.
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
  • Compatible with downstream HLA assays
The EasySep™ HLA T Cell Enrichment Kit is designed to isolate T cells from fresh or previously frozen peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-T cells and dextran-coated magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use ofMore Product Information