EasySep™ Mouse SCA1 Positive Selection Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18756RF 18756
STMC18756RFEA 1106 EUR
STMC18756RF STMC18756
EasySep™ Mouse SCA1 Positive Selection Kits
Media voor celscheiding
Immunomagnetic positive selection of SCA1+ cells (e.g. hematopoietic stem/progenitor cells) from mouse bone marrow.
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
The EasySep™ Mouse SCA1 Positive Selection Kit is designed to isolate SCA1+ cells from single-cell suspensions of bone marrow by positive selection. The cocktail contains a combination of PE-labelled antibodies that is directed against SCA1, which are then recognized by antibody complexes that are directed against PE and dextran. labelled cells are bound to magnetic particles and separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.
This kit replaces the EasySep™ Mouse SCA1 Biotin Positive Selection Kit for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').