EasySep™ Human CD15 Positive Selection Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18651RF 18651
STMC18651RFEA 829 EUR
STMC18651RF STMC18651
EasySep™ Human CD15 Positive Selection Kits
Media voor celscheiding
Immunomagnetic positive selection of human CD15+ cells (e.g. Myeloid cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human CD15 Positive Selection Kit is designed to isolate CD15+ cells from ammonium chloride lysed blood or other nucleated cell suspensions by positive selection. Desired cells are targeted with antibody complexes recognising CD15 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD15 antigen is expressed on monocytes and granulocytes.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').