EasySep™ Rat Custom Positive Selection Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18609 18609RF
STMC18609EA 0 EUR
STMC18609 STMC18609RF
EasySep™ Rat Custom Positive Selection Kits
Media voor celscheiding
Immunomagnetic positive selection of rat cells.
The EasySep™ Rat Custom Positive Selection Kit is designed to isolate any rat cells from fresh splenocytes or single cell suspension of other tissues by positive selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').