EasySep™ Human Custom Positive Selection Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
18309 18309RF
STMC18309EA 0 EUR
STMC18309 STMC18309RF
EasySep™ Human Custom Positive Selection Kits
Media voor celscheiding
Immunomagnetic positive selection of human cells.
The EasySep™ Human Custom Positive Selection Kit is designed to isolate any cell type of interest from fresh or previously frozen peripheral blood mononuclear cells, buffy coat or whole blood by positive selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team for more information.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').