EasySep™ Human Progenitor Cell Enrichment Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17936 17936RF
STMC17936EA 594 EUR
STMC17936 STMC17936RF
EasySep™ Human Progenitor Cell Enrichment Kit II
Media voor celscheiding
Immunomagnetic enrichment of untouched human hematopoietic progenitor cells.
  • Fast, easy-to-use, and column-free procedure
  • Up to 95% purity
  • Untouched, viable cells
The EasySep™ Human Progenitor Cell Enrichment Kit II is designed to isolate hematopoietic progenitor cells from fresh cord blood and other cell preparations by negative selection. The EasySep™ procedure involves labelling unwanted cells with antibody complexes and magnetic particles. The magnetically labelled cells are separated from the desired cells by using an EasySep™ magnet and simplyMore Product Information