EasySep™ Human CD45 Depletion Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17898 17898RF
STMC17898EA 880 EUR
STMC17898 STMC17898RF
EasySep™ Human CD45 Depletion Kit II
Media voor celscheiding
Immunomagnetic depletion of human CD45+ cells.
  • Fast, easy-to-use and column-free
  • Up to 4 log depletion of CD45+ cells
  • Isolated cells are untouched
The EasySep™ Human CD45 Depletion Kit II is designed to deplete CD45+ cells from fresh or previously frozen human peripheral blood mononuclear cells. Unwanted cells are targeted with antibody complexes recognising CD45 and magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube while desired cells are poured off. The CD45 antigen is expressed on all hematopoietic cells except erythrocytes and platelets.
This product replaces the EasySep™ Human CD45 Depletion Kit for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').