EasySep™ Human Myeloid Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17893 17893RF
STMC17893EA 668 EUR
STMC17893 STMC17893RF
EasySep™ Human Myeloid Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of human CD33+ and CD66+ cells (e.g. myeloid cells).
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ Human Myeloid Positive Selection Kit II is designed to isolate CD33+ and CD66b+ cells from ammonium chloride lysed blood or bone marrow by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD33, CD66b and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 and CD66b antigens are expressed on myeloid progenitors monocytes and granulocytes.
This product replaces the EasySep™ Human Myeloid Positive Selection Kit for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').