EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17887 17887RF
STMC17887EA 930 EUR
STMC17887 STMC17887RF
EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of human CD138+ cells from whole blood and bone marrow (e.g. normal or malignant plasma cells for multiple myeloma research).
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human Whole Blood and Bone Marrow CD138 Positive Selection Kit is designed to isolate CD138+ to learn how to process bone marrow samples and isolate CD138+ plasma cells to increase sensitivity in FISH testing for multiple myeloma. You can also browse our Frequently Asked Questions about isolating CD138+ plasma cells to improve FISHMore Product Information