EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17885 17885RF
STMC17885EA 889 EUR
STMC17885 STMC17885RF
EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kits
Media voor celscheiding
Column-free immunomagnetic positive selection of human CD33+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kit is designed to isolate highly purified myeloid (including CD14+ and CD66b+) cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD33 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD33 antigen is expressed on myeloid progenitor cells, granulocytes and monocytes.
This product replaces the EasySep™ Human Whole Blood CD33 Positive Selection Kit and EasySep™ HLA Whole Blood CD33 Positive Selection Kit for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').