EasySep™ Human CD2 Positive Selection Kit II
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17883 17883RF
STMC17883EA 777 EUR
STMC17883 STMC17883RF
EasySep™ Human CD2 Positive Selection Kit II
Media voor celscheiding
Immunomagnetic positive selection of CD2+ cells.
  • Fast and easy-to-use
  • Up to 97% purity
  • No columns required
The EasySep™ Human CD2 Positive Selection Kit II is designed to isolate CD2+ cells from fresh or previously frozen human peripheral blood mononuclear cells by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD2 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD2 antigen is expressed on T cells and NK cells.
This product replaces the EasySep™ Human CD2 Positive Selection Kit for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').