EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kits
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
17878 17878RF
STMC17878EA 777 EUR
STMC17878 STMC17878RF
EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kits
Media voor celscheiding
Column-free immunomagnetic positive selection of human CD14+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Buffy Coat CD14 Positive Selection Kit is designed to isolate CD14+ cells from freshly prepared buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD14 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD14 antigen is strongly expressed on monocytes and is weakly expressed on granulocytes. It is also expressed on most tissue macrophages.
This product replaces the EasySep™ Human Buffy Coat CD14 Positive Selection Kit for even faster cell isolations.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').