Collagenase C, ACF
Leverancier: STEMCELL Technologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
07442 07443
STMC07442EA 146 EUR
STMC07442 STMC07443
Collagenase C, ACF
Celcultuursupplementen en -additieven
Animal component-free collagenase for the digestion of native collagen fibrils.
  • Obtain cleaner cultures with an animal component-free solution
Use Collagenase C, Animal Component-Free (ACF) to digest native collagen fibrils in connective tissues and prevent the introduction of animal-derived pathogens. This enzymatic preparation is sourced from Clostridium histolyticum and is obtained from cultures free of animal-derived materials. Collagenase preparations contain the activity of several proteases including: collagenase, caseinase, clostripain, and trypsin (Kessler and Yaron). Similar to Collagenase Type IV, Collagenase C, ACF contains low levels of tryptic activity, limiting damage to membrane proteins and receptors.
Opgelet: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').