Transfermembranen Nytran® N, Whatman™
Leverancier: Whatman products (Cytiva)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
10416096 10416196
732-4045EA 636 EUR
732-4045 732-4056
Transfermembranen Nytran® N, Whatman™
Transfermembranen
The Nytran® Nylon (Nytran® N) membrane is ideal for applications that require a lower charge. It is designed for Southern and Northern blotting as well as colony and plaque lifts and Dot or Slot Blots. Nytran® N is compatible with isotopic detection methods.
  • Uitstekende signaal-ruisverhouding
  • Het membraan wordt uniform aan beide zijden van de ondersteunende matrix aangebracht en is daarom perfect symmetrisch. Hierdoor ligt het vlak zonder aan de randen op te krullen
  • Uniforme poriëngrootte en -verdeling
  • Leverbaar met poriëngrootte 0,2 µm en 0,45 µm, voor optimale retentie van oligo's en relatief grote DNA-fragmenten
Nytran N membrane allows for excellent signal-to-noise ratios. The membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling. Nytran N membrane is a highly consistent membrane with uniform pore size and distribution. It is available in 0,2 and 0,45 µm pore sizes for optimal retention of oligos and larger DNA fragments.
Whatman Nytran nylon membranes are suitable for applications that require a lower charge. They are designed for Southern and Northern blotting as well as colony and plaque lifts and dot-/slot-blots. Nytran N is compatible with isotopic and nonisotopic detection methods.