VWR®, LB media, pre-mixed
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J106-1KGEA 236 EUR
J106-1KG J106-10PK J106-2KG
VWR®, LB media, pre-mixed
Media voor microbiologie Gedehydrateerde media voor microbiologie
Lysogeny broth (LB) is a nutritionally rich medium used for the propagation and maintenance of E. coli. It is also widely used for the preparation of plasmid DNA and recombinant proteins.
  • Save time using pre-mixed media
  • Homogenous and high quality powders
Formulation of 10 g tryptone, 10 g NaCl, and 5 g yeast extract (25 g powder total) per L of liquid broth.
The instructions included on the label state to prepare LB broth by dissolving 25 g powder per liter in deionized water.
Leveringsinformatie: Individual ReadyPack™ pouches prepare 1 L of media.